missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11393837
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a polymerization catalyst, a chlorinating agent, an intermediate in chemical synthesis, also a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
PubChem CID 24294
IUPAC Name antimony pentachloride
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 79°C (22mmHg)
Melting Point 2.8°C
Physical Form Liquid
Quantity 1 g
UN Number UN1730
Merck Index 14,695
Solubility Information Soluble in water (react),chloroform,hydrochloric acid,and carbon tetrachloride.
Formula Weight 299.02
Percent Purity 99.999%
Odor Pungent
Refractive Index 1.601
Sensitivity Moisture sensitive
Density 2.336 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(V) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.