missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(V) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific™
Flame retardant
41.00 €
Chemical Identifiers
CAS | 1314-60-9 |
---|---|
Molecular Formula | O5Sb2 |
Molecular Weight (g/mol) | 323.515 |
MDL Number | MFCD00011216 |
InChI Key | LJCFOYOSGPHIOO-UHFFFAOYSA-N |
Synonym | antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40 |
PubChem CID | 14813 |
IUPAC Name | (dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane |
SMILES | O=[Sb](=O)O[Sb](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359267
|
Thermo Scientific Alfa Aesar
040128.36 |
500 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1314-60-9 | |
323.515 | |
LJCFOYOSGPHIOO-UHFFFAOYSA-N | |
14813 | |
O=[Sb](=O)O[Sb](=O)=O |
O5Sb2 | |
MFCD00011216 | |
antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40 | |
(dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane |
Specifications
1314-60-9 | |
O5Sb2 | |
antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40 | |
O=[Sb](=O)O[Sb](=O)=O | |
323.515 | |
323.5 | |
Odorless |
Particles in Liquid | |
MFCD00011216 | |
LJCFOYOSGPHIOO-UHFFFAOYSA-N | |
(dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane | |
14813 | |
30% in H2O | |
Antimony(V) oxide, Colloidal dispersion |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 215-237-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures