Learn More
Invitrogen™ Anza™ 5-Pack Starter Kit
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN3004
Product Code. 15521620
  • 253.00 € / Each
Explore more special offers
Includes: Anza™ 11 EcoRI, Anza™ 5 BamHI, Anza™ 1 NotI, Anza™ 16 HindIII, Anza™ 12 XbaI, Anza™ T4 DNA Ligase Master Mix, Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer,
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA substrates
  • Complete digestion in 15 min
  • Includes 5 common restriction enzymes
  • Complete with Anza T4 DNA Ligase Master Mix and Anza Alkaline Phosphatase
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
• 100 μL Anza T4 DNA Ligase Master Mix
• 20 μL Anza Alkaline Phosphatase
• 500 μL Anza Buffer (10X)
• 500 μL Anza Red Buffer (10X)
Store at -5 to -30°C.
Methylation Sensitivities Varies by Enzyme
Anza™ 10X Red Buffer, Anza™ 10X Buffer
1 kit
Traditional Cloning
Approved for shipment on Wet or Dry Ice
HindIII, NotI, Alkaline Phosphatase, XbaI, T4 DNA Ligase, BamHI, EcoRI
No
Starter Kit
Anza™