missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dbo sieges AP6 Polyurethane Stool
Stool made of polyurethane steel for use in workshops and garages.
160.00 € - 9575.43 €
Specifications
Adjustable | Height |
---|---|
Frame Material | Steel |
Quantity | 1 Ea. |
Seat Material | Polyurethane |
Warranty | 5 Years |
Product Code | Brand | Seating Height | With Casters | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Seating Height | With Casters | Price | Quantity & Availability | |||||
15724308
|
dbo sieges
AP6B1D1NE2 |
Standard | Yes |
167.00 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
15734308
|
dbo sieges
AP6B1D1NE3 |
Standard | No |
160.00 €
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
15744308
|
dbo sieges
AP6B2D1NE2 |
Medium | Yes |
171.00 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
15754308
|
dbo sieges
AP6B2D1NE3 |
Medium | No |
9575.43 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
15764308
|
dbo sieges
AP6B3D1NE3 |
High | No |
170.00 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
Description
- Available in different central cylinder heights.
- Seat of molded polyurethane on wooden structure with inserts.
- Height adjustment.
- Five-spoke base of injected nylon and fiberglass.
- Available with caster or slider feet.
Specifications
Height | |
1 Ea. | |
5 Years | |
No | |
Stool |
Steel | |
Polyurethane | |
No | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
dbo sieges AP6 Polyurethane Stool