Learn More
Thermo Scientific™ ApaI (10 U/μL)
Cut at GGGCC^C sites with ApaI restriction enzyme, which functions best at 37°C in B buffer (Isoschizomers: PspOMI).
75.00 € - 105.00 €
Specifications
Concentration 10 U/μL
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10130690
View Documents
Thermo Scientific™
ER1411
3000 U
75.00 €
3000 units
10550841
View Documents
Thermo Scientific™
ER1415
5,000 units
105.00 €
5000 units
Description
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications
Specifications
10 U/μL