Learn More
A&D Weighing Apollo GF-A Series Precision Balance
A&D Apollo Analytical Series improves productivity and efficiency of your workflows with user-friendly and innovative features. It is designed to provide reliable results across a range of environments and applications.
1495.00 € - 2505.00 €
Specifications
Certifications/Compliance | GLP/GMP/GCP/ISO |
---|---|
Connection Type | RS-232 C and USB |
Display Type | Reverse-backlit LCD |
Material | Stainless Steel |
Electrical Requirements | 30 VA to adapter (12 V, 1 A) |
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
16601112
|
A&D Weighing
GF-203A |
220 g | 0.001 g |
1655.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16611112
|
A&D Weighing
GF-3002A |
3200 g | 0.01 g |
1770.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16621112
|
A&D Weighing
GF-303A |
320 g | 0.001 g |
1770.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16631102
|
A&D Weighing
GF-10001A |
10,200 g | 0.1 g |
1880.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16631112
|
A&D Weighing
GF-4002A |
4200 g | 0.01 g |
1845.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16641102
|
A&D Weighing
GF-10002A |
10,200 g | 0.01 g |
2505.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16641112
|
A&D Weighing
GF-403A |
420 g | 0.001 g |
1845.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16651102
|
A&D Weighing
GF-1003A |
1100 g | 0.001 g |
2280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16651112
|
A&D Weighing
GF-6001A |
6200 g | 0.1 g |
1725.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16661102
|
A&D Weighing
GF-1202A |
1220 g | 0.01 g |
1495.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16661112
|
A&D Weighing
GF-6002A |
6200 g | 0.01 g |
2190.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16671102
|
A&D Weighing
GF-123A |
122 g | 0.001 g |
1495.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16671112
|
A&D Weighing
GF-603A |
620 g | 0.001 g |
2160.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16681102
|
A&D Weighing
GF-1603A |
1620 g | 0.001 g |
2435.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16691102
|
A&D Weighing
GF-2002A |
2200 g | 0.01 g |
1655.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
- Newly developed Smart SHS weighing sensor featuring ECL technology
- APA (Auto Precision Assessment) – Automatic assessment of the balance performance, calculation and display of Standard Deviation (repeatability) at the point of use without the need for calibration weights, can be checked daily via a key press
- Automatic Minimum Weight Calculation and setting via APA function
- Input minimum weight automatically via APA or manually using weights
- Visual Minimum Weight alert
- UAC (User Access Control) – Administrator and Operator password access to ensure data integrity
- Calibration via One-touch, Interval, pre-set times (3) or automatically in response to ambient temperature changes (GX-AE/A)
- ISD (Patented Impact Shock Detection) – 4 level shock detection with visual and audible feedback, maintain the performance of your investment through correct use of the balance, ideal in automated loading applications and ideal for operator guidance
- FRD (Flow Rate Display) – Direct display of flow rate of a sample in g/s, g/m, g/h or volume flow rate in mL/s, mL/m, mL/h
- Up to 10 sample densities can be saved to memory
- Use in conjunction with the flow Rate Comparator to keep flow rates within designated limits
- SCF (Statistical Calculation Function) – Display and print various statistical results on the balance display or output to a printer or PC
- Standard RS232 and Dual Mode USB interface. USB can be configured for quick mode (connect to PC without the need for software installation) or bi-directional mode
- UFC (Universal Flexi Coms) – configurable data output, ideal for connection to data acquisition systems or barcode label printers etc.
- Gross/Net/Tare data output, Tare memory, Underhook, Capacity Indicator
- Large selection of Options and Accessories
- Built-in fanless ionizer with Quick Ion™ technology and external IR switch (GX-AE)
Especificaciones
GLP/GMP/GCP/ISO | |
Reverse-backlit LCD | |
30 VA to adapter (12 V, 1 A) |
RS-232 C and USB | |
Stainless Steel |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto