missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apoptosis Activator 2, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Description | Orange |
| Quantity | 5 mg |
| Formula Weight | 306.14g/mol |
| Sensitivity | Light sensitive |
| Product Type | Apoptosis Reagent |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Activates apoptosis in cells, shows selectivity for cancer cells against normal cell lines |
| InChI Key | KGRJPLRFGLMQMV-UHFFFAOYSA-N |
| SMILES | ClC1=CC=C(CN2C(=O)C(=O)C3=CC=CC=C23)C=C1Cl |
| IUPAC Name | 1-[(3,4-dichlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione |
| Show More |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur