Learn More
Appendix IX Organophosphorous/Nitrogen Pesticides and Herbicides, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand:  SPEX Certiprep 8270-IXJ
Product Code. 15275856
  • 333.00 € / 1mL
Explore more special offers
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Show More
Specifications
Specifications
Appendix IX Organophosphorus/Nitrogen Pest & Herb
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Method 8270
1 mL
For US EPA Methods 8270, 8270C
Acetone
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Appendix IX Organophosphorous/Nitrogen Pesticides and Herbicides, SPEX CertiPrep™ > 1mL; 2mL amber prescored glass ampule
Product intended for laboratory use only