missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Professional™ Appliance lid
Appliance lid DE-CS7-85 for the top cover on the machine, 70 cm deep, 90 cm wide (PG 8583 CD)
Brand: Miele Professional™ 10222780
337.37 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
This item is not returnable.
View return policy
Description
Accessory for washer-disinfector PG 8583 CDSpecifications
Stainless Steel | |
Stainless Steel | |
900 mm |
Cover | |
Washer-disinfector PG 8583 CD | |
15 mm |