missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq™ 360 DNA Polymerase
GREENER_CHOICE

The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.

Brand:  Applied Biosystems™ 4398828

 View more versions of this product

Product Code. 10055024

  • 538.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: AmpliTaq 360 DNA Polymerase is supplied at 5U/μL with 10X AmpliTaq 360 PCR Buffer, 25mM M2Cl2, and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq 360 Buffer, 25mM MgCl2, and 360 GC Enhancer is supplied with each 250U of enzyme. Storage Buffer Enzyme is supplied in 20mM Tris, pH 9.0, 100mM KCl, 0.1mM EDTA, 1mM DTT, 0.5% (w/v) Tween 20, 50% (v/v) glycerol. Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.
This item is not returnable. View return policy

Description

Description

The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. Compared to the original AmpliTaq DNA Polymerase, AmpliTaq 360 DNA Polymerase is purified by an additional proprietary separation process that reduces contaminating bacterial DNA sequences from the enzyme preparation. This ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and, when combined with the proprietary AmpliTaq 360 Buffer Kit, promotes the amplification of a variety of templates, including those from bacterial and human genomes.

Features

  • Optimized for the broadest range of targets, from everyday to challenging
  • Unmatched sensitivity and yield
  • Market-leading GC enhancer for robust amplification of GC-rich sequences
  • Achieves the highest quality sequencing data

Notes

  • DreamTaq DNA Polymerase is recommended for superior PCR performance.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• AmpliTaq 360 DNA Polymerase (5 U/μL), 200 μL
• 10X AmpliTaq 360 PCR Buffer, 4 x 1.5 mL
• 25 mM MgCl2, 4 x 1.5 mL
• 360 GC Enhancer, 4 x 1.5 mL

Store at -15°C to -25°C.
Product is guaranteed up to 12 months as indicated by the expiration date.
Medium
Standard
Sustainable packaging
1000 Units
Standard PCR
1X
800 Reactions
Separate Components
DNA
Stand-alone enzyme
AmpliTaq 360 DNA Polymerase
5' - 3'
DNA Polymerase
Room Temp or Wet Ice
5 U/μL
No
3'-A
5 kb or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.