Learn More
Applied Biosystems™ AmpliTaq™ 360 DNA Polymerase
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.
Brand:  Applied Biosystems™ 4398893
 View more versions of this product
Product Code. 10279094
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: AmpliTaq 360 DNA Polymerase is supplied at 5U/μL with 10X AmpliTaq 360 PCR Buffer, 25mM M2Cl2, and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq 360 Buffer, 25mM MgCl2, and 360 GC Enhancer is supplied with each 250U of enzyme. Storage Buffer Enzyme is supplied in 20mM Tris, pH 9.0, 100mM KCl, 0.1mM EDTA, 1mM DTT, 0.5% (w/v) Tween 20, 50% (v/v) glycerol. Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.
Description
Description
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. Compared to the original AmpliTaq DNA Polymerase, AmpliTaq 360 DNA Polymerase is purified by an additional proprietary separation process that reduces contaminating bacterial DNA sequences from the enzyme preparation. This ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and, when combined with the proprietary AmpliTaq 360 Buffer Kit, promotes the amplification of a variety of templates, including those from bacterial and human genomes.
Features
  • Optimized for the broadest range of targets, from everyday to challenging
  • Unmatched sensitivity and yield
  • Market-leading GC enhancer for robust amplification of GC-rich sequences
  • Achieves the highest quality sequencing data
Notes
  • DreamTaq DNA Polymerase is recommended for superior PCR performance.
Order Info
Approved for shipment at Room Temperature or on Wet Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 10X AmpliTaq 360 PCR Buffer, 1.5 mL for each 250 U of enzyme
• 25 mM MgCl2, 1.5 mL for each 250 U of enzyme
• 360 GC Enhancer, 1.5 mL for each 250 U of enzyme
Storage Buffer Enzyme is supplied in 20 mM Tris (pH 9.0), 100 mM KCl, 0.1 mM EDTA, 1 mM DTT, 0.5% (w/v), Tween 20, 50% (v/v) glycerol
Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli
All solutions are prepared in deionized, filter-sterilized water.
When stored at -15°C to -25°C, this product maintains performance at least until the expiration date on the tube label. Before use, thaw frozen components to room temperature and mix well.
Product is guaranteed up to 12 months as indicated by the expiration date.
High
5' - 3'
5 U/μL
3'-A
5 kb or less
Stand-alone enzyme
AmpliTaq Gold 360 DNA Polymerase
3000 Units
1X
Separate Components
DNA