missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea)

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

56.00 € - 321.00 €

Specifications

Form Liquid
Product Line GeneAmp™
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Concentration 10 mM
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10381565
View Documents
Applied Biosystems™
N8080260
1 mL
56.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10157054
View Documents
Applied Biosystems™
4303441
6 x 1 mL
321.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use

Specifications

Specifications

Liquid
Dry Ice
10 mM
GeneAmp™
Standard PCR
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.