Learn More
Applied Biosystems™ GeneAmp™ Fast PCR Master Mix (2X), with or without protocol
The GeneAmp Fast PCR Master Mix, with its sensitive, pre-mixed hot-start chemistries, reduces PCR reaction time to as little as 25 minutes when used with the Veriti 96-Well Thermal Cycler.
246.00 € - 305.00 €
Specifications
Format | Tube |
---|---|
GC-Rich PCR Performance | Medium |
Polymerase | AmpliTaq DNA Polymerase |
Reaction Speed | Fast |
Exonuclease Activity | 5' - 3' |
Includes: The GeneAmp Fast PCR Master Mix (2X) contains two tubes (1.25mL each) sufficient for 125 20μL PCR reactions. The Fast PCR Master Mix can also be ordered with a protocol.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10229534
|
Applied Biosystems™
4362070 |
250 Reactions (with protocol) | N/A | ||||||
10526745
|
Applied Biosystems™
4359187 |
250 Reactions (withithout protocol) |
305.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Applied Biosystems GeneAmp Fast PCR Master Mix reduces PCR reaction time to as little as 25 minutes when used with the Veriti 96-well thermal cycler. The sensitive, premixed hot-start chemistries of this master mix enable quick experiment setup and by generating greater than 90% 'A' overhang, the cloning process is further simplified.
GeneAmp Fast PCR Master Mix may be used to detect a single copy of a gene in 10 ng of human genomic DNA or as little as 10 copies of an HIV target.
Features
- Quick reaction setup with premixed reagents
- Gene detection with as little as 10 ng of human genomic DNA
- Reproducible, specific, and sensitive results
Notes
- The Applied Biosystems Fast PCR System, consisting of the premixed GeneAmp Fast PCR Master Mix, specially designed microplates and tubes, and the Veriti 96-well thermal cycler (0.1 mL or 0.2 mL), considerably reduces PCR reaction times.
- GeneAmp Fast PCR Master Mix is provided at a convienient 2X concentration with only the addition of DNA template and primers required.
Order Info
Shipping Condition: Dry ice
Specifications
Tube | |
AmpliTaq DNA Polymerase | |
5' - 3' | |
2X | |
Built-In Hot Start | |
3'-A | |
2 kb or less |
Medium | |
Fast | |
Fast PCR Master Mix | |
1X | |
250 Reactions | |
SuperMix or Master Mix | |
0.1 mL or 0.2 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.