missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ Thermal Isolation Frame

Balances thermal uniformity across 0.5mL GeneAmp PCR System 9700

Brand:  Applied Biosystems™ 4308927

Product Code. 10035704

  • 87.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: 8 GeneAmp Thermal Isolation Frames.
This item is not returnable. View return policy

Description

Description

When used with the 0.5mL GeneAmp reaction tubes, the GeneAmp Thermal Isolation Frame creates a proper thermal and physical seal between the heated cover and the reaction tubes.

  • Sturdy,reinforced fiberglass ensures precise PCR results
  • Convenient tool protects precious samples
  • Maintain Sample Temperature Uniformity
  • During PCR, it is critical that thermal cycler lid does not compress sample tubes and heat samples capriciously
  • Fits snugly around samples to ensure reproducible results
  • Designed for Convenience
  • Fits around thermal cycler sample block
  • When thermal cycler lid is closed, frame ensures even contact between lid and tubes
  • Leave frame on thermal cycler for continual back-to-back runs

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
GeneAmp
Thermal Isolation Frame
PCR
Room Temperature
Contains 8 GeneAmp™ Thermal Isolation Frames. Store at room temperature.
Black
GeneAmp 9700
8 frames
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.