Learn More
Applied Biosystems™ High-Capacity cDNA Reverse Transcription Kit
The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction.
538.00 € - 4135.00 €
Specifications
Format | Kit |
---|---|
Reaction Speed | 120 min. |
Technique | Reverse Transcription |
Quantity | Each |
Reverse Transcriptase | MultiScribe |
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
10400745
|
Applied Biosystems™
4368814 |
Kit only | 200 Reactions |
538.00 €
200 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249814
|
Applied Biosystems™
4374966 |
Kit with Rnase Inhibitor | 200 Reactions |
974.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10186954
|
Applied Biosystems™
4368813 |
Kit only | 1,000 Reactions |
2165.00 €
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743647
|
Applied Biosystems™
4374967 |
Kit with Rnase Inhibitor | 1,000 Reactions |
4135.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction. Both types of kit deliver extremely high-quality, single-stranded cDNA from 0.02–2 μg total RNA. Reactions can be scaled up to 100 μL to generate 10 μg of cDNA from a single reaction.
The High Capacity cDNA Reverse Transcription Kit with RNase Inhibitor includes the recombinant enzyme used to prevent degradation of RNA templates and lacks DNA endonuclease activity for better product yield.
In addition to their use in RT-PCR, this kit may be used in traditional PCR, microarrays, and for the generation of cDNA archives.
Features of the High Capacity cDNA Reverse Transcription Kit include:
• Linear target amplification for real-time PCR
• Higher yields and precision than other cDNA synthesis kits at a fraction of the cost
• 10-fold greater dynamic range than other kits
Extensively tested with a variety of templates
Quantitative first-strand synthesis of all RNA species is achieved with the use of the random primers. This kit has been extensively tested with a variety of RNA templates, including GC- and AU-rich targets and RNAs expressed at low levels. cRNA can also be efficiently generated from in vitro transcription of cDNA.
Informations sur la commande
Shipping Condition: Dry ice
Spécification
Kit | |
Reverse Transcription | |
MultiScribe | |
Dry Ice | |
First-Strand cDNA | |
Reverse Transcription |
120 min. | |
Each | |
Yes | |
50 U/μL | |
Separate components | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.