missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MeltDoctor™ HRM Calibration Standard (20X)

Contains a DNA template and primers

Brand:  Applied Biosystems™ 4425562

279.90 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10126904

  • 622.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 1 tube containing 1mL of a 20X solution of DNA template and primers which, when mixed with PCR reagents and dye, can be used to prepare a calibration plate for thermal and optical High Resolution Melt (HRM).
This item is not returnable. View return policy

Description

Description

Can be used to prepare a plate for thermal and optical High Resolution Melt (HRM) calibration.

  • Applied Biosystems MeltDoctor HRM Calibration Standard contains a DNA template and primers which, when mixed with PCR reagents and dye, can be used to prepare a plate for thermal and optical High Resolution Melt calibration

Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, High Resolution Melting (HRM) Analysis, Methylation Analysis, Methylation-Sensitive HRM Analysis, PCR & Real-Time PCR, RNAi, Epigenetics & Noncoding RNA Research, Real Time PCR (qPCR)

Order Info

Wet ice shipping; store at -15° to -25°C; guaranteed minimum shelf life is 60 days

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tube
HRM (High Resolution Melting) Dye
MeltDoctor™
Real Time PCR (qPCR)
1 mL
20 X
1 tube containing 1 mL of a 20X solution of DNA template and primers which, when mixed with PCR reagents and dye, can be used to prepare a calibration plate for thermal and optical High Resolution Melt (HRM).
Store at -15°C to -25°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
7500 Fast System, 7900HT Fast System
HRM Calibration Standard
Wet Ice
Thermal Calibration, Optic Calibration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.