missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Tray/Retainer Set for Veriti™ Systems

Engineered for efficient sample processing

Brand:  Applied Biosystems™ 4381850

124.45 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.


Product Code. 10764617

  • 132.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 10 MicroAmp 96-Well Tray/Retainer Sets.
This item is not returnable. View return policy

Description

Description

Specifically designed to snugly hold MicroAmp 8-tube strips (0.2mL) and MicroAmp reaction tubes without caps (0.2mL) in a 96-well format for processing on the Veriti 96-Well Thermal Cycler.

  • Designed specifically for MicroAmp8-Tubes Strip (0.2mL) and MicroAmp reaction tubes without Caps (0.2mL)<
  • Manufactured to high specification, ensuring quality product

  • Allows to organize tubes while adding sample and mix
  • Reduces spillage and tube mix-ups by securing and ordering tubes on lab bench
  • When samples are ready, tray of tubes are put directly on block of Veriti 96-Well Thermal Cycler
  • Eliminates hassle of transferring tubes and provides thermal contact necessary for thermal cycling

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
MicroAmp
Tray/Retainer Set
PCR
10 Trays
Contains 10 MicroAmp™ 96-Well Tray⁄Retainer Sets. Store at room temperature.
Blue
SimpliAmp™ Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Thermal Cycler, ProFlex™ PCR System
96-Well Plates
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.