missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Tray

Specifically designed to snugly hold up to 96 MicroAmpReaction tubes with caps

Brand:  Applied Biosystems™ N8010541

Additional Details : Weight : 0.29000kg

Product Code. 10147494

  • 112.00 € / Each
In Stock
to see stock.
Add to basket

Includes: 10 MicroAmp 96-Well Bases.

Description

Description

The MicroAmp 96-well tray is required for more efficient sample processing on Applied Biosystems96-well thermal cyclers.

  • Engineered for efficient sample processing
  • Designed specifically for MicroAmp reaction tubes with Caps
  • Manufactured to high specification, ensuring quality product

  • Allows to organize tubes while adding sample and mix
  • Reduces spillage and tube mix-ups by securing and ordering your tubes on lab bench
  • When samples are ready, tray of tubes are directly on block of thermal cycler
  • Eliminates hassle of transferring tubes
  • Provides thermal contact necessary for thermal cycling

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Certified DNA/RNase Free
MicroAmp
Tray
PCR
10 Trays
Contains 10 MicroAmp™ 96-Well Trays. Store at room temperature.
Natural
GeneAmp 9700, 2720 Thermal Cycler
96-Well Plates
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers