Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Fast Clear Reaction Plates with Barcode
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
Brand: Applied Biosystems™ 4483494
Description
- Optimized for performance with Applied Biosystems instruments
- Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
- Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
- Great mechanical stability and flatness for complete workflow needs
- Use of polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability
- Ideal choice for use with automation/robotics and multi-instrument experiments (Like thermal cycler, real-time thermal cycler, genetic analyzer)
- Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
- Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity
- Platform: 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7500 Fast Dx System, 7500 Fast System, 7900HT Fast System, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Fast Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Fast System
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications
Includes Barcode | |
Store at room temperature. | |
MicroAmp | |
Microplate | |
Semi | |
0.2 mL | |
0.1 mL | |
Fast |
Certified DNA/RNase Free | |
500 Plates | |
Clear, Optical | |
Thermal Cycler, Genetic Analyzer, DNA Analyzer | |
Polycarbonate, Polypropylene | |
96 | |
500 plates |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
I28A2Q724 | Certificate of Analysis | 25/08/2022 | 4483494_LITL18 PROMO,4483494 |
I28A2Q722 | Certificate of Analysis | 25/08/2022 | 4483494_LITL18 PROMO,4483494 |
I28A2Q624 | Certificate of Analysis | 25/08/2022 | 4483494_LITL18 PROMO,4483494 |
I28A2Q622 | Certificate of Analysis | 25/08/2022 | 4483494_LITL18 PROMO,4483494 |
I28A2Q621 | Certificate of Analysis | 25/08/2022 | 4483494_LITL18 PROMO,4483494 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.