missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Fast 96-Well Tray

Designed to snugly hold MicroAmp Fast reaction tubes with cap (0.1mL) and MicroAmp Fast 8-tube strips

Brand:  Applied Biosystems™ 4358305

Product Code. 10478844

  • 141.00 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: 10 MicroAmp Fast 96-Well Trays.
This item is not returnable. View return policy

Description

Description

The MicroAmp fast 96-well tray is required for more efficient sample processing on the Applied Biosystems9800 Fast Thermal Cycler.

  • Organizes fast reaction tubes or strips
    Provides thermal contact necessary for fast PCR
    Manufactured to high specification, ensuring quality production

  • Allows organize tubes while adding sample and mix
  • Reduces spillage and tube mix-ups by securing and ordering tubes on lab bench
  • When samples are ready, tray of tubes are put directly on block of 9800 Fast Thermal Cycler
  • Eliminates hassle of transferring tubes and provides thermal contact necessary for efficient and accurate thermal cycling

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
MicroAmp
9800 Fast Thermal Cycler
96-Well Plates
Room Temperature
Contains 10 MicroAmp™ Fast 96-Well Trays. Store at room temperature.
Tray
PCR
10 Trays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers