missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Splash-Free 96-Well Base

Used for centrifugation

Brand:  Applied Biosystems™ 4312063

162.45 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.


Product Code. 10659655

  • 171.00 € / Pack of 10
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: 10 MicroAmp Splash Free 96-Well Bases.
This item is not returnable. View return policy

Description

Description

  • Designed to hold MicroAmp reaction tubes in MicroAmp tray/retainer sets or 96-well reaction plates for convenient pre-PCR and post-PCR sample processing
  • Uniform Results
  • Use with instruments that have heated sample covers to eliminate condensation and need for mineral oil overlay
  • Proprietary MicroAmp reaction tubes are essential for high-performance PCR to ensure maximum temperature and amplification uniformity for samples

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
MicroAmp
96-Well Base
PCR
10 Bases
Contains 10 MicroAmp™ Splash Free 96-Well Bases. Store at room temperature.
Black
7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOne™, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx
96-Well Plates
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.