missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiScribe™ Reverse Transcriptase

MultiScribe Reverse Transcriptase is a recombinant moloney murine leukemia virus (rMoMuLV) reverse transcriptase optimized for TaqMan-based assays.

Brand:  Invitrogen™ 4311235

Product Code. 10320165

  • 192.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: MultiScribe™ Reverse Transcriptase (100μL at 50U/μL).
This item is not returnable. View return policy

Description

Description

MultiScribe Reverse Transcriptase is a recombinant moloney murine leukemia virus (rMoMuLV) reverse transcriptase optimized for TaqMan-based assays. MultiScribe Reverse Transcriptase is similar to MuLV reverse transcriptase but differs in its recommended usage. This recombinant RNA–dependent, DNA polymerase uses single-stranded RNA as a template in the presence of a primer to synthesize a complementary DNA (cDNA) strand.

Features of MultiScribe Reverse Transcriptase include:
• Compatible with one-step and two-step RT-PCR reactions, which provides flexibility in assay setup
• Individual component design provides versatility in assay setup
• 5,000 U is supplied in 100 μL vials (50 U/μL)
• Reaction temperature up to 52°C

Applications
• PCR
• RT-PCR
• Reverse Transcription

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• MultiScribe Reverse Transcriptase, 100 μL (50 U/μL)

Store at –20°C.

Reverse Transcription
100 μL
Reduced
RT-PCR
First-Strand cDNA
Reverse Transcription
RNA
Stand-alone Enzyme
37°C
MultiScribe
Dry Ice
50 U/μL
Separate components
Up to 7 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.