missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format)

These 0.2mL PCR tubes with caps are provided in an 8-strip format and guaranteed RNase- and DNase-free.

Brand:  Applied Biosystems™ AM12230

Product Code. 10505225

  • 363.00 € / Pack of 125

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

These 0.2mL PCR tubes with caps are provided in an 8-strip format and guaranteed RNase- and DNase-free. Tubes and caps design allows caps to seal securely with minimal pressure. Polypropylene tubes are autoclave-safe.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
125 Strips
Polypropylene
RNase-Free, DNase-Free
Room Temperature
Autoclavable
Tube
0.2 mL
Thermal Cyclers
Store at room temperature.
Standard
8-Strip Cap
Yes
PCR Tube and Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.