missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ PreAmp Master Mix Kit
TaqMan™ PreAmp Master Mix Kit contains TaqMan™ PreAmp Master Mix and TaqMan™ Gene Expression Master Mix.
Brand: Applied Biosystems™ 4384267
This item is not returnable.
View return policy
Description
- Amplify cDNA targets equally without introducing bias
- Analyze mRNA from any precious sample such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE)
- Stretch as little as 1ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression Assays
- Analyze up to 100 gene expression targets with minimal hands-on time
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Contains 2X TaqMan PreAmp Master Mix (1 mL), TaqMan™ Gene Expression Master Mix (5 mL), and TaqMan™ PreAmp Master Mix Kit Quick Reference Card. Store master mixes at 2-8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
High | |
AmpliTaq Gold DNA Polymerase | |
7300 System, 7500 Fast System, 7500 System, 7900HT Fast System | |
TaqMan™ | |
40 reactions | |
Built-In Hot Start |
Primer-probe | |
qPCR | |
Fast or Standard | |
No ROX | |
PreAmp Master Mix Kit | |
200 Reactions | |
cDNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction