missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ True Allele™ PCR Premix

Applied Biosystems True Allele PCR Premix is a PCR master mix that contains AmpliTaq Gold DNA Polymerase, dNTPs, MgCl2, and buffer.

Brand:  Applied Biosystems™ 403061

1667.25 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10732687

  • 2565.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 10 tubes, each containing 1.8mL of True Allele™ PCR Premix, enough to perform 2000 PCR amplifications of 15μL each.
This item is not returnable. View return policy

Description

Description

Applied Biosystems True Allele PCR Premix is a PCR master mix that contains AmpliTaq Gold DNA Polymerase, dNTPs, MgCl2, and buffer. It is intended for use with the ABI PRISM Linkage Mapping Set. True Allele PCR Premix has been optimized for strong, specific PCR amplification of microsatellite markers.

Accessory products

ABI PRISM Linkage Mapping Set

Notes

See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 tubes of True Allele PCR Premix, 1.8 mL each

Store at -20°C.
AmpliTaq DNA Polymerase
5' - 3'
1 kit
Hot-start PCR
1X
2000 Reactions
5 kb or less
Tube
Standard
PCR Premix
Dry Ice
5 U/μL
Built-In Hot Start
Separate Components
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.