Learn More
Thermo Scientific™ Aqua-Tec™ Water Preservation Cell
Protect your valuable samples from water borne contaminants without the use of harsh chemicals.
Brand: Thermo Scientific™ 50109579
432.17 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
The Thermo Scientific™ Aqua-Tec™ Water Preservation Cell is a revolutionary way to protect your valuable samples from water borne contaminants without the use of harsh chemicals. Simply place the 3.7 in. (9.4 cm) cell into the water reservoir of any CO2 incubator or water bath. Aqua-Tec prevents contamination from most types of bacteria, molds, and fungi for up to six months.
- Designed and tested specifically for laboratory use in CO2 incubators and water baths
- Proven effective for up to 6 months against bacteria, molds, and fungi
- Safe to handle– requires no special disposal protocol
- Self-regulates to maintain effectiveness regardless of water level
- Easy-to-use in equipment from any manufacturer
Compliance
US EPA Registration 91025-1-95500
Specifications
Germany (N-86509), Switzerland (CHZN2388), Ireland (PCS No. 100694) | |
Elemental Silver | |
Pack of 4 cells | |
11 mm |
CO2 Incubators and Water Baths | |
Water Preservation Cell | |
94 mm | |
24 mm |
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
This product is only available in the UK, Ireland, Germany, France, Austria, Switzerland, Sweden, Finland, and Iceland