missing translation for 'onlineSavingsMsg'
Learn More

3M™ Nomad™ Aqua Textile Drop Down Matting Roll, Series 85

Advanced water removal and retention properties keep floors cleaner and safer, for an extended period of time. Traditional, deep cut pile and coir matting allows dirt and moisture to sit on the surface and be tracked into the building.

Brand:  3M™ Nomad™ 7000032563

 View more versions of this product

Product Code. 17118580

  • 3845.00 € / 1 roll
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High performing textile entrance mat, with advanced water removal and retention properties
  • Stops 80% of dirt and moisture at the door
  • Entrance mat for high traffic applications (1500 to 5000 crossings per day)
  • Phthalate-free, slip-resistant PVC backing
  • Excellent appearance retention
  • Long lasting and easy to clean
  • Available as drop down mats or in rolls
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 x 2 m
Nomad™ Aqua, Series 85
Gray
2 m
Heavy traffic areas
7.5 mm
10 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.