missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Aquadur™ Test Strips

Determine water hardness quickly, easily and reliably. The Aquadur™ test strips provide a convenient dip and read procedure and a clear colour changes from green to red ensures reliable results within seconds.

33.75 €

Specifications

Certifications/Compliance CE, 93/42 / EEC
Format Strip
For Use With (Application) Determination of water hardness
Quantity 100 Pack
Test Time 20 sec.
View More Specs

Products 2
Product Code Brand Quantity Graduations Price Quantity & Availability  
Product Code Brand Quantity Graduations Price Quantity & Availability  
10632381
View Documents
Macherey-Nagel™
91201
100 Pack <3 > 5 > 10 > 15 <20 > 25°d
33.75 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10028220
View Documents
Macherey-Nagel™
91220
100 Pack <3 > 4 > 7 > 14 <21°d
33.75 €
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

  • Quick and easy determination of water hardness.
  • The convenient dip and read procedure and the clear colour changes from green to red ensure reliable results within seconds.
  • Water hardness test strips come in a wide variety of different gradations and packaging options for your individual needs.
Specifications

Specifications

CE, 93/42 / EEC
Determination of water hardness
20 sec.
Strip
100 Pack
Aquadur Test Strips
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.