missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Complete 1, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Single-component reagent; Pyridine free

200.00 €

Specifications

Purity or Free of Statement Pyridine free
Chemical Name or Material Aqualine™ Complete 1
Color Colorless
Name Note Karl Fischer Reagent
CAS 111-90-0, 288-32-4, 7446-09-5, 7553-56-2
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10538810
View Documents
Fisher Chemical
K/1900/15
1 L Amber glass bottle
200.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 1.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
Specifications

Specifications

Pyridine free
Colorless
111-90-0, 288-32-4, 7446-09-5, 7553-56-2
Liquid
Aqualine™ Complete 1
Karl Fischer Reagent
1.2mg of water/mL min.
Solution
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Irritating to skin. Irritating to eyes. Possible risk of harm to the unborn child.
NFPA
Health: 1

Signal Word: DANGER!

Product Identifier
  • Aqualine Complete 1

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyA§ thoroughly after handling.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.