missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aqualine™ Complete 2, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Color:
Brown
Colorless
Unit Size:
1 L
2.5 L
Description
- Single-component volumetric reagent for high water content analysis
- Water equivalent: 2.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free
Specifications
Specifications
| Chemical Name or Material | Aqualine™ Complete 2 |
| Name Note | Karl Fischer Reagent |
| Color | Colorless |
| CAS | 7446-09-5, 288-32-4, 111-90-0, 7553-56-2 |
| Physical Form | Liquid |
| Quantity | 2.5 L |
| Molecular Formula | Solution |
| MDL Number | 81734 |
| Packaging | Amber glass bottle |
Safety and Handling
Product Identifier
- Aqualine]TS Complete 2
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H360D-May damage the unborn child.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- H315-Causes skin irritation.
- P201-Obtain special instructions before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto