missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
83.65 € - 710.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433813
|
Fisher Chemical
K/2250R/08 |
500 mL |
83.65 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10092042
|
Fisher Chemical
K/2250R/15 |
1 L |
710.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Single-component volumetric reagent for high water content analysis of aldehydes and ketones
- Water equivalent: 5.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free
Specifications
Amber glass bottle | |
Liquid | |
81734 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Aqualine Complete 5K
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical™