missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Brand: Fisher Chemical K/2110/17
137.45 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Solvent for samples with high hydrocarbon content
- Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis
Specifications
Aqualine™ Solvent CM | |
Pyridine free | |
67-66-3, 288-32-4, 7446-09-5 | |
Liquid | |
81734 |
Karl Fischer Reagent | |
2.5 L | |
Amber glass bottle | |
Pass Test |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Aqualine Solvent CM
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
- P308+P313-IF exposed or concerned: Get medical advice/attention.