missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ AquaPhenol™
Water saturated phenol for DNA/RNA extractions
344.00 € - 506.68 €
Specifications
Content And Storage | Store at +4°C. Store under nitrogen. Protect from light. |
---|---|
Formulation | Water saturated Phenol |
pH | 4.5 - 5 (Lit.) |
Research Category | Molecular Biology |
Form | Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11466960
|
MP Biomedicals™
11AQUAPH01 |
250 mL |
344.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11476960
|
MP Biomedicals™
11AQUAPH02 |
4 x 250 mL | N/A | ||||||
Description
Phenol is used as a reagent in chemical analysis. It is used in the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes. It is commonly used in tissue preservation and DNA/RNA extraction with chloroform.Specifications
Store at +4°C. Store under nitrogen. Protect from light. | |
4.5 - 5 (Lit.) | |
Solution |
Water saturated Phenol | |
Molecular Biology |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- AquaPhenol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Phenol, ultra pure
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.