missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AR-C 69931 Tetrasodium Salt
Low molecular weight, organic compound that may help regulate biological process.
Brand: Tocris Bioscience™ 5720/1
This item is not returnable.
View return policy
Description
AR-C 69931 tetrasodium salt is a highly potent P2Y12 antagonist (IC50 = 0.4 nM). Inhibits ADP-induced human platelet aggregation ex vivo. Reduces thrombus weight and delays thrombus formation in a model of carotid artery thrombosis in vivo.Specifications
Antagonist | |
163706-36-3 | |
864.29g/mol | |
Store at -20°C | |
1 mg |
P2Y Receptor | |
C17H21Cl2F3N5Na4O12P3S2 | |
Cangrelor tetrasodium salt | |
P2Y Receptor Antagonists |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction