missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ AR-GLAS™ Stirring Rods

Product Code. 10283821 Shop All Brand Products
Click to view available options
Length (Metric):
200 mm
250 mm
300 mm
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10283821

Brand: BRAND™ 135810

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Constructed of AR-GLAS™. Tubes are fused closely at both ends.

  • Constructed of AR-GLAS™
  • Tubes are fused closely at both ends.
  • Supplied in packs of 50
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6 mm
Length (Metric) 200 mm
Material Glass
Quantity 50/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.