missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)
ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)
120.00 € - 375.00 €
Specifications
Detection Method | Fluorescence |
---|---|
Diameter (Metric) | 6 μm |
Product Type | Compensation Beads |
Format | Tube |
For Use With (Equipment) | Flow Cytometer |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10093324
|
Invitrogen™
A10346 |
ArC™ Amine Reactive Compensation Bead Kit, 1 Kit, 100 rxns, 6μm Diameter, 6μm Bead Diameter, Fluorescent Detection Method, Flow Cytometry Compensation Beads Calibration Type, Surface Stain Configuration, Any lasers (UV to 633/635) Flow Cytomet | 1 Kit |
375.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15563052
|
Invitrogen™
A10628 |
ArC™ Amine Reactive Compensation Bead Kit, 1 Kit, 25 Tests rxns, 6μm Diameter, 6μm Bead Diameter, Fluorescent Detection Method, Flow Cytometry Compensation Beads Calibration Type, Surface Stain Configuration, Any lasers (UV to 633/635) Flow Cy | 1 Kit |
120.00 €
1 set |
In Stock
Log in to see stock. |
||||
Description
ArC™ Amine Reactive Compensation Bead Kit is a bead-based compensation tool specifically optimized for use with LIVE/DEAD™ Fixable dead cell stain kits.View a selection guide for all fixable viability dyes for flow cytometry.
Ready-to-use control
Unlike other methods that require sample preparation to make compensation controls using cellular material, the ArC™ Amine Reactive Compensation Bead Kit is ready to use out of the dropper bottle. The kit includes two types of specially modified polystyrene microspheres that have been optimized to react with the amine-reactive dye in the LIVE/DEAD™ Fixable dead cell stain kits. The ArC™ reactive beads (Component A) bind any of the amine-reactive dyes and provide a positive signal; the ArC™ negative beads (Component B) have no reactivity and provide a negative compensation control. This ready-to-use product preserves the original sample material for experiments; precious sample is not used for the creation of compensation controls.
Accurate and consistent results
Unlike biological controls, which can have variable expression levels depending upon disease or development state, the Arc™ beads provide a bright, consistent signal (see figure below). Consistency in the staining pattern of compensation controls is important for obtaining the right compensation value. If the signal varies with the control material used, the compensation value can be affected, leading to erroneous results.
How it works
One drop of beads is added to a test tube containing LIVE/DEAD™ Fixable dead cell stain. After incubation with amine-reactive dye, the beads are washed in staining buffer, a drop of negative control beads is added (if required), and the beads are resuspended and analyzed by flow cytometry. The two components provide a distinct positive bead that is at least 50-fold brighter than the negative beads, which can be used to set compensation.
Other compensation products
Besides the ArC™ Amine Reactive Compensation Beads, Life Technologies also offers the AbC™ Anti-Mouse Bead Kit for capture of mouse anti-human antibodies and AbC™ Anti-Rat/Hamster Bead Kit for capture of rat anti-mouse or hamster anti-mouse antibodies.
Specifications
Fluorescence | |
Compensation Beads | |
Flow Cytometer | |
Room Temperature |
6 μm | |
Tube | |
1 Kit | |
Surface |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title