Learn More
Tocris Bioscience™ ARCC 4 negative control
Negative control for ARCC 4 (Cat. No. 7254)
Brand:  Tocris Bioscience™ 7255/5
364.57 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000884
  • 471.00 € / 5mg
Explore more special offers
Description
Description
ARCC 4 negative control is a negative control for ARCC 4 (Cat. No. 7254). Binds androgen receptors (AR) without inducing degradation.
Specifications
Specifications
(2 S,4 S)-1-(-2-(2-(4-((4 '-(3-(4-Cyano-3-(trifluoromethyl)phenyl)-5,5-dimethyl-4-oxo-2-thioxoimidazolidin-1-yl)-[1,1 '-biphenyl]-4-yl)oxy)butoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxami
5 mg
98%
2156588-26-8
C53H56F3N7O7S2
Active Degrader Negative Controls