Learn More
Thermo Scientific™ ARCTIC A10B Refrigerated Circulators
A10B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from -10° to +100°C and 30L maximum bath volume.
Brand: Thermo Scientific™ 1524101
Description
The Thermo Scientific™ A10B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from -10° to +100°C and maximum bath volume of 30 liters. Configure your refrigerated bath with a Thermo Scientific STANDARD or ADVANCED controller that meets your specific application needs.
Refrigerated baths are used primarily for controlling the temperature of samples placed within them, so the work area dimensions will be important specifications to evaluate. These units can also circulate for a limited set of refrigerated applications.- Robust stainless-steel baths
- Thermostat can be indexed 90° on all sides of the bath for easy viewing
- External circulation set included for plumbing connection M16×1, 0.25 NPT male, 0.375in. O.D.
- Three-side ventilation concept: up to two sides can be blocked with full refrigeration performance and no degradation of equipment
- Draining port at the front
- Integrated handles
- CFC-free refrigerant
Includes:
Control cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation, and 6ft. power cord.
Recommended for:
- Calibration
- Bioreactors
- Rotary Evaporators
- Condensers
- Sample/Material Testing
- Sample/Material Preparation
Specifications
0.6 to 1.0 cu. ft. | |
17 to 30 L | |
250 W | |
-10°C to +100°C | |
1/NFL | |
SC100 | |
35.9 in. | |
91.3 cm | |
16.9 in. | |
42.9 cm | |
18.5 in. | |
7.9 in. | |
97.9 lb. | |
230 V 50 Hz | |
50 Hz |
SC100 | |
CE/ROHS/WEEE | |
17 L/min. | |
300 mbar/4.35 psi | |
0.02°C | |
35.9 x 16.9 x 18.5 in. (91.3 x 42.9 x 47.1 cm) | |
13.4 in. | |
36.5 cm | |
11.7 in. | |
29.7 cm | |
47.1 cm | |
20 cm | |
44.5 kg | |
230 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.