Learn More
Sartorius Arium™ Advance L Reverse Osmosis System
Arium™ Advance L Reverse Osmosis System for the production of pure water.
Brand:  Sartorius H2O-RO-H-100
16490.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16837296
  • 18440.00 € / Each
Explore more special offers
Description
Description
  • The Arium™ Advance L laboratory water systems are designed for a reverse osmosis water requirement of up to 200 L/hr
  • Combined with the integrated 100 or 200 liter storage tank the system provides the ideal volume of pre-treated feed water for your laboratory.
  • The system can simultaneous feed several laboratory instruments at once, thanks to the powerful distributor pump.
  • Its compact, square shape reduces space requirements to a minimum.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 L
200 L/hr
Water System
0.05 MΩ*cm
Potable Tap Water
Reverse Osmosis (Type 3)
0.25 in.