missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Arium™ Bagtank w/o Integrated Pump

Sartorius Stedim Biotech™ Arium™ Bagtank w/o Integrated Pump uses a closed bagtank system to protect your prepared pure water from secondary contamination.

Brand:  Sartorius Stedim Biotech™ H2O-AOV-100-W

 View more versions of this product

Product Code. 15923020

  • 1760.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stores pure water in the innovative, closed Ariumbagtank system.
  • Protects pure water from secondary contamination.
  • Enables consistent water quality over an extended period, thereby providing for long-term, reproducible results.
  • In contrast to typical water tanks, the Arium bag provides a high degree of user safety and saves time by the elimination of complex sanitization processes with chemicals.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 L
Arium Comfort, Arium Advance
Gravity
EU plug, bagtank (w/o bags), installation kit
85.2cm × 51.4cm × 58.7cm
Pure Water Storage System, 100 L
Pure water storage
47 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers