Learn More
Sartorius arium™ comfort I ASTM 1, 3 Combined Water purification Systems with UV Lamp



Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
Brand: Sartorius H2O-I-1-UV-B
8840.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Unique cartridge kit for production of highest quality ultrapure water
- Innovative iJust technology optimizes product water quality
- Touch display with intuitive menu navigation
- Compact design with SD card slot
- Reproducible and reliable results
- Arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
- Available as bench-top system for every laboratory workstation
- Wall-mounted with integrated wall bracket
Benefits
- Optimized, economic water consumption
- Premium product-water quality at all times
- Guaranteed longer life of downstream ultrapure water systems

Specifications
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) | |
<0.001 EU/mL with a terminal ultrafiltration cartridge | |
No | |
8 L/hr. | |
No | |
≤2 ppb | |
2.0 to 6.9 bar | |
HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, Photometry, Buffer solutions, Feed water for laboratory devices, such as autoclaves, glassware washers etc | |
Wall | |
Adsorption by spherical activated carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration | |
17 in. | |
19.7 in. | |
18.7 in. | |
68.3 lb. (Operating); 50.7 lb. (Empty) | |
100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.) |
120 L | |
3/8 in. | |
Tap | |
Type 1 and 3 | |
18.2 Mohm.cm | |
Water Purification System | |
2°C to 30°C | |
80% | |
<1mL | |
<1 NTU | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
Approx. 31 kg (Operating); 23 kg (Empty) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.