missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ mini Compact Ultrapure Water System
BIOPHARMA

The arium mini compact ultrapure water system developed to provied ASTM Type 1 water quality. The standard version arium mini is independent of permanently installed water tap. Cat. No. 14-559-041 offers an integrated UV lamp (185 ∣ 254nm) to reduce TOC down to < 5 ppb.

Brand:  Sartorius H2O-MM-UV-T

3677.29 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15565153

  • 4410.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simple bag exchange in less than 5min.
  • No use of cleaning agents needed
  • Optimized purity, due to closed system, avoiding permanent biofilm
  • Intuitive menu navigation with high quality glass touch-display, developed through customer survey
  • Small footprint 28cm (11 in.) width
  • Volume dispense from 50mL
  • For general and analytical applications at a daily need<10L
  • Feed water supplied to system via provided container, filled with pre-treated water
TRUSTED_SUSTAINABILITY
Specifications

Specifications

arium™ mini with UV Lamp Ultrapure Water System
1 L/min.
18.2 Mohm.cm at 25°C
Water Purification System
Benchtop
Distilled, DI, RO, EDI
Type 1
<5 ppb
Integrated UV lamp (185/254nm), Power cord
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers