missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ mini UV lamp (185 ∣ 254nm)

For use with arium mini and arium mini plus Ultrapure Water Systems

Brand:  Sartorius H2O-CEL1

246.85 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 15515153

  • 286.00 € / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ultrapure water, free of TOC
  • Horizontal installation, optimized temperature gradient
  • Effective breakdown of organic compounds
  • Prevents the growth of microorganisms
  • Easy replacement
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UV Lamp
arium mini and arium mini plus Ultrapure Water Systems
185/254 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius arium™ mini UV lamp (185 ∣ 254nm) > UV Lamp

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.