missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10922-1-AP-20UL
This item is not returnable.
View return policy
Description
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97kDa (beta-form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.Specifications
ARP2 | |
Polyclonal | |
Unconjugated | |
ACTR2 | |
Actin like protein 2, Actin related protein 2, ARP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10097, 289820, 66713 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61160, P61161, Q5M7U6 | |
Actr2 | |
ARP2 Fusion Protein Ag1359 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |