missing translation for 'onlineSavingsMsg'
Learn More

Notrax Arrow Trax™ Non-Directional Chevron Patterned Entrance Mat

Notrax Entrance Mat - The needle punched fibers are durable and crush-resistant for extreme grime retention making it suitable for heavy traffic areas. Dense vinyl backing is moisture resistant and slip resistant to keep the mat in place.

Brand:  Notrax 118S5110CH

 View more versions of this product

Product Code. 17701948

  • 289.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The non-directional chevron pattern of the Arrow Trax™ ensures a very aggressive scraping action making it one of our most popular carpet mats offering tremendous value
  • The needle punched fibers are durable and crush-resistant for extreme grime retention making it suitable for heavy traffic areas
  • Dense vinyl backing is moisture resistant and slip resistant to keep the mat in place
Specifications

Specifications

Charcoal
300 cm
Industrial and Commercial Matting
118 in.
Polyester
130 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers