missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Arsenic trioxide is used as a precursor to forestry products, in colorless glass production, and in electronics. And it is also used as pigments, and preservatives for hides and wood, in the semiconductor industry to create light emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 465°C (sublimation) |
Melting Point | 312.3°C |
Physical Form | Powder |
Quantity | 2 g |
UN Number | UN1561 |
Merck Index | 14,804 |
Solubility Information | Soluble in hot water,dilute acids and alkalies. Insoluble in alcohol,chloroform,and ether,. |
Formula Weight | 197.84 |
Percent Purity | 99.996% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'