missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

69.45 € - 244.22 €

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389508
View Documents
Thermo Scientific Alfa Aesar
043340.06
5 g
69.45 €
5g
Please to purchase this item. Need a web account? Register with us today!
11379508
View Documents
Thermo Scientific Alfa Aesar
043340.14
25 g
244.22 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For vacuum deposition.Arsenic(III) sulfide is used to make infrared-transmitting glass, pyrotechnics, pigments, linoleum, electrical semiconductors and photoconductors. It acts as a reducing agent and serves as a tanning agent. It is also used for depilating hides and veterinary medication. Further, it is used as an inorganic photoresist to fabricate 3-D nanostructures. In addition to this, it is used in the treatment for acute promyelocytic leukemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-33-9
246.02
CUGMJFZCCDSABL-UHFFFAOYSA-N
[S--].[S--].[S--].[As+3].[As+3]
As2S3
MFCD00003435
diarsenic(3+) trisulfanediide

Specifications

707°C
1303-33-9
Fused Pieces
As2S3
Insoluble in water.
[S--].[S--].[S--].[As+3].[As+3]
246.02
Like rotten eggs
Arsenic(III) sulfide
300°C
Yellow
99.99% (Metals basis)
MFCD00003435
CUGMJFZCCDSABL-UHFFFAOYSA-N
diarsenic(3+) trisulfanediide
246.03
3.43 g/cm3
Videos
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 215-117-4

missing translation for 'rtecsNumber' : CG2638000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.