missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

166.83 € - 615.00 €

Chemical Identifiers

CAS 12044-54-1
Molecular Formula As2Te3
Molecular Weight (g/mol) 532.643
MDL Number MFCD00014167
InChI Key GTIUFDICMGTSPM-UHFFFAOYSA-N
Synonym arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
PubChem CID 15770922
SMILES [As](=[Te])[Te][As]=[Te]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11368848
SDS View Documents
Thermo Scientific Alfa Aesar
041708.06
5 g
166.83 €
5g
Please to purchase this item. Need a web account? Register with us today!
11358848
SDS View Documents
Thermo Scientific Alfa Aesar
041708.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12044-54-1
532.643
GTIUFDICMGTSPM-UHFFFAOYSA-N
15770922
As2Te3
MFCD00014167
arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
[As](=[Te])[Te][As]=[Te]

Specifications

381°C
Powder
MFCD00014167
arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
[As](=[Te])[Te][As]=[Te]
15770922
99.999%
6.50 g/mL
Arsenic(III) telluride
12044-54-1
As2Te3
UN1557
GTIUFDICMGTSPM-UHFFFAOYSA-N
532.643
532.64
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 234-955-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.