missing translation for 'onlineSavingsMsg'
Learn More
Arsenic lump, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11396577

Arsenic lump, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11396577
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11396577

Brand: Thermo Scientific Alfa Aesar 000590.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Assay Percent Range 99.99% (metals basis)
Quantity 100 g
Chemical Name or Material Arsenic lump

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW23K020 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number000590.22,000590.14
Lot NumberP26K015 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number000590.22,000590.14
Lot NumberU29J004 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number000590.22,000590.14
Lot NumberT24J076 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number000590.22,000590.14
Lot NumberS09J007 Certificate TypeCertificate of Analysis Date11/07/2023 Catalog Number000590.22,000590.14,000590

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic lump, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.