missing translation for 'onlineSavingsMsg'
Learn More

Arsenic lump, 99.999% (metals basis), Thermo Scientific Chemicals

197.99 € - 597.98 €

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386577
View Documents
Thermo Scientific Alfa Aesar
000590.14
25 g
197.99 €
25g
Please to purchase this item. Need a web account? Register with us today!
11396577
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
000590.22
100 g
597.98 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Identifiants chimiques

7440-38-2
74.92
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
CHEBI:27563
[As]

Spécification

7440-38-2
As
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
[As]
74.92
CHEBI:27563
99.99% (metals basis)
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
Arsenic lump
Vidéos
Sécurité et traitement

Sécurité et traitement

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 213-148-6

missing translation for 'rtecsNumber' : CG0525000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Identifiant produit
  • Arsenic lump

Mention d'avertissement
  • Danger

Catégorie de danger
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Mention de danger
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H410-Very toxic to aquatic life with long lasting effects.

Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

FDS
Documents

Documents

Certificats

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.